Assay Details
| Gene Symbol: CD72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015117489.2 | XP_014972975.2 | 2144 | 69 | ||
| XM_015117490.2 | XP_014972976.2 | 2147 | 69 | |||
| XM_015117491.2 | XP_014972977.2 | 440 | 69 | |||
| XM_028835080.1 | XP_028690913.1 | 421 | 69 | |||
| GenBank mRNA | JU496937.1 | - | 284 | 69 | ||
More Information
Set Membership: |
|

CD72
Genomic Map