Assay Details
| Gene Symbol: ABHD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121647.2 | XP_014977133.1 | 970 | 98 | ||
| XM_015121648.2 | XP_014977134.1 | 966 | 98 | |||
| XM_015121649.2 | XP_014977135.1 | 710 | 98 | |||
| XM_028837961.1 | XP_028693794.1 | 966 | 98 | |||
| XM_028837962.1 | XP_028693795.1 | 966 | 98 | |||
| GenBank mRNA | JU322351.1 | - | 920 | 98 | ||
| JV044764.1 | - | 917 | 98 | |||
More Information
Set Membership: |
|

ABHD3
Genomic Map