Assay Details
| Gene Symbol: TOX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126117.2 | XP_014981603.2 | 3154 | 81 | ||
| XM_015126118.2 | XP_014981604.2 | 2897 | 81 | |||
| GenBank mRNA | JU341906.1 | - | 818 | 81 | ||
More Information
Set Membership: |
|

TOX3
Genomic Map