Assay Details
| Gene Symbol: ZNF775 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015135263.2 | XP_014990749.2 | 103 | 70 | ||
| GenBank mRNA | JU484985.1 | - | 91 | 70 | ||
| JV614086.1 | - | 82 | 70 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015135263.2 transcripts.
Set Membership: |
|

ZNF775
Genomic Map