Assay Details
| Gene Symbol: EBPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121087.2 | XP_014976573.2 | 336 | 96 | ||
| GenBank mRNA | JV484637.1 | - | 73 | 96 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EBPL| Gene Symbol: EBPL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121087.2 | XP_014976573.2 | 336 | 96 | ||
| GenBank mRNA | JV484637.1 | - | 73 | 96 | ||
Set Membership: |
|