Assay Details
| Gene Symbol: C16H17orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU325023.1 | - | 926 | 93 | ||
| JV714576.1 | - | 947 | 93 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C16H17orf100| Gene Symbol: C16H17orf100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU325023.1 | - | 926 | 93 | ||
| JV714576.1 | - | 947 | 93 | |||
Set Membership: |
|