Assay Details
| Gene Symbol: PABPC1L2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV580414.1 | - | 122 | 81 | ||
| Gene Symbol: LOC106995243 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028841958.1 | XP_028697791.1 | 1464 | 81 | ||
More Information
Set Membership: |
|

PABPC1L2B
Genomic Map