Assay Details
| Gene Symbol: ALKAL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028832030.1 | XP_028687863.1 | 560 | 67 | ||
| GenBank mRNA | JV058622.1 | - | 211 | 67 | ||
| JV648964.1 | - | 333 | 67 | |||
More Information
Set Membership: |
|

ALKAL2
Genomic Map