Assay Details
| Gene Symbol: ATP11AUN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028837420.1 | XP_028693253.1 | 92 | |||
| GenBank mRNA | JV651302.1 | - | 115 | 92 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP11AUN| Gene Symbol: ATP11AUN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028837420.1 | XP_028693253.1 | 92 | |||
| GenBank mRNA | JV651302.1 | - | 115 | 92 | ||
Set Membership: |
|