Assay Details
| Gene Symbol: RFX8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112972.2 | XP_014968458.1 | 1490 | 74 | ||
| XM_015112973.2 | XP_014968459.1 | 1566 | 74 | |||
| XM_015112974.2 | XP_014968460.1 | 1276 | 74 | |||
| XM_015112975.2 | XP_014968461.1 | 1634 | 74 | |||
| XM_015112976.2 | XP_014968462.1 | 1417 | 74 | |||
| XM_028832246.1 | XP_028688079.1 | 1381 | 74 | |||
| XM_028832249.1 | XP_028688082.1 | 1742 | 74 | |||
| GenBank mRNA | JV660436.1 | - | 119 | 74 | ||
More Information
Set Membership: |
|

RFX8
Genomic Map