Assay Details
| Gene Symbol: ZNF773 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028839883.1 | XP_028695716.1 | 1208 | 104 | ||
| XM_028839884.1 | XP_028695717.1 | 1205 | 104 | |||
| GenBank mRNA | JU328036.1 | - | 222 | 104 | ||
More Information
Set Membership: |
|

ZNF773
Genomic Map