Assay Details
| Gene Symbol: MYL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149055.2 | XP_015004541.1 | 513 | 88 | ||
| GenBank mRNA | JU332496.1 | - | 428 | 88 | ||
| JU473970.1 | - | 390 | 88 | |||
| JV045898.1 | - | 414 | 88 | |||
More Information
Set Membership: |
|

MYL9
Genomic Map