Assay Details
| Gene Symbol: PARP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015146183.2 | XP_015001669.2 | 1548 | 87 | ||
| XM_015146184.2 | XP_015001670.2 | 1548 | 87 | |||
| GenBank mRNA | JU495098.1 | - | 1537 | 87 | ||
| JV663795.1 | - | 1476 | 87 | |||
| Gene Symbol: LOC114680148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028852301.1 | XP_028708134.1 | 1573 | 87 | ||

PARP10
Genomic Map