Assay Details
| Gene Symbol: LOC720345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015149884.2 | XP_015005370.2 | 1302 | 79 | ||
| XM_015149885.2 | XP_015005371.2 | 1302 | 79 | |||
| XM_015149886.2 | XP_015005372.2 | 1302 | 79 | |||
| XM_015149887.2 | XP_015005373.2 | 1071 | 79 | |||
| XM_028828573.1 | XP_028684406.1 | 1302 | 79 | |||
| XM_028828574.1 | XP_028684407.1 | 1302 | 79 | |||
| GenBank mRNA | JU473383.1 | - | 1121 | 79 | ||
More Information
Set Membership: |
|

LOC720345
Genomic Map