Assay Details
| Gene Symbol: DMWD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028839491.1 | XP_028695324.1 | 692 | 78 | ||
| XM_028839492.1 | XP_028695325.1 | 705 | 78 | |||
| XM_028839493.1 | XP_028695326.1 | 685 | 78 | |||
| GenBank mRNA | JV046347.1 | - | 634 | 78 | ||
| JV667355.1 | - | 630 | 78 | |||
| JV708584.1 | - | 452 | 78 | |||
More Information
Set Membership: |
|

DMWD
Genomic Map