Assay Details
| Gene Symbol: PRICKLE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105952.4 | XP_001105952.2 | 1925 | 60 | ||
| GenBank mRNA | JV651312.1 | - | 315 | 60 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRICKLE3| Gene Symbol: PRICKLE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001105952.4 | XP_001105952.2 | 1925 | 60 | ||
| GenBank mRNA | JV651312.1 | - | 315 | 60 | ||
Set Membership: |
|