Assay Details
| Gene Symbol: MAFK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133158.2 | XP_014988644.1 | 188 | 98 | ||
| GenBank mRNA | JU336415.1 | - | 63 | 98 | ||
| JU477231.1 | - | 87 | 98 | |||
| JV601233.1 | - | 161 | 98 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015133158.2 transcripts.
Set Membership: |
|

MAFK
Genomic Map