Assay Details
| Gene Symbol: MAFK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133157.2 | XP_014988643.1 | 2089 | 66 | ||
| XM_015133158.2 | XP_014988644.1 | 270 | 66 | |||
| XM_028845485.1 | XP_028701318.1 | 218 | 66 | |||
| XM_028845486.1 | XP_028701319.1 | 292 | 66 | |||
| GenBank mRNA | JU336415.1 | - | 145 | 66 | ||
| JU477231.1 | - | 169 | 66 | |||
| JV601233.1 | - | 243 | 66 | |||
More Information
Set Membership: |
|

MAFK
Genomic Map