Assay Details
| Gene Symbol: MRTFA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150425.2 | XP_015005911.2 | 1610 | 70 | ||
| XM_015150426.2 | XP_015005912.2 | 1690 | 70 | |||
| XM_015150427.2 | XP_015005913.2 | 1628 | 70 | |||
| XM_028827670.1 | XP_028683503.1 | 1596 | 70 | |||
| GenBank mRNA | JU476551.1 | - | 1574 | 70 | ||
| JV047766.1 | - | 1501 | 70 | |||
| JV667432.1 | - | 1656 | 70 | |||
More Information
Set Membership: |
|

MRTFA
Genomic Map