Assay Details
| Gene Symbol: MRTFA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015150425.2 | XP_015005911.2 | 2837 | 112 | ||
| XM_015150426.2 | XP_015005912.2 | 2917 | 112 | |||
| XM_015150427.2 | XP_015005913.2 | 2855 | 112 | |||
| GenBank mRNA | JU476551.1 | - | 2801 | 112 | ||
| JV047766.1 | - | 2728 | 112 | |||
| JV667432.1 | - | 2883 | 112 | |||
More Information
Set Membership: |
|

MRTFA
Genomic Map