Assay Details
| Gene Symbol: TBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028827984.1 | XP_028683817.1 | 952 | 96 | ||
| GenBank mRNA | JV069724.1 | - | 706 | 96 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TBX1| Gene Symbol: TBX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028827984.1 | XP_028683817.1 | 952 | 96 | ||
| GenBank mRNA | JV069724.1 | - | 706 | 96 | ||
Set Membership: |
|