Assay Details
| Gene Symbol: GEMIN6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015112154.2 | XP_014967640.1 | 159 | 72 | ||
| GenBank mRNA | JV661739.1 | - | 54 | 72 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015112154.2 transcripts.
Set Membership: |
|

GEMIN6
Genomic Map