Assay Details
| Gene Symbol: ZNF480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015124757.2 | XP_014980243.2 | 124 | |||
| XM_015124758.2 | XP_014980244.2 | 124 | ||||
| XM_015124759.2 | XP_014980245.2 | 124 | ||||
| XM_028839412.1 | XP_028695245.1 | 124 | ||||
| GenBank mRNA | JV707624.1 | - | 379 | 124 | ||
More Information
Set Membership: |
|

ZNF480
Genomic Map