Assay Details
| Gene Symbol: PCDH11X | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127772.2 | XP_014983258.1 | 3566 | 65 | ||
| XM_028842285.1 | XP_028698118.1 | 3408 | 65 | |||
| XM_028842286.1 | XP_028698119.1 | 3408 | 65 | |||
| GenBank mRNA | JU334032.1 | - | 3193 | 65 | ||
| JU334033.1 | - | 3193 | 65 | |||
More Information
Set Membership: |
|

PCDH11X
Genomic Map