Assay Details
| Gene Symbol: USP51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015127550.2 | XP_014983036.1 | 787 | 65 | ||
| XM_028841918.1 | XP_028697751.1 | 873 | 65 | |||
| XM_028841919.1 | XP_028697752.1 | 753 | 65 | |||
| XM_028841920.1 | XP_028697753.1 | 636 | 65 | |||
| XM_028841921.1 | XP_028697754.1 | 873 | 65 | |||
| GenBank mRNA | JV572813.1 | - | 221 | 65 | ||
More Information
Set Membership: |
|

USP51
Genomic Map