Assay Details
| Gene Symbol: CCNJL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001084076.4 | XP_001084076.3 | 861 | 78 | ||
| XM_028849883.1 | XP_028705716.1 | 785 | 78 | |||
| XM_028849884.1 | XP_028705717.1 | 743 | 78 | |||
| XM_028849885.1 | XP_028705718.1 | 962 | 78 | |||
| XM_028849886.1 | XP_028705719.1 | 957 | 78 | |||
| GenBank mRNA | JV071259.1 | - | 296 | 78 | ||
| JV661391.1 | - | 310 | 78 | |||
More Information
Set Membership: |
|

CCNJL
Genomic Map