Assay Details
| Gene Symbol: DOP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015136840.2 | XP_014992326.2 | 321 | 107 | ||
| XM_015136841.2 | XP_014992327.2 | 322 | 107 | |||
| XM_015136843.2 | XP_014992329.2 | 283 | 107 | |||
| XM_015136844.2 | XP_014992330.2 | 321 | 107 | |||
| XM_015136847.2 | XP_014992333.2 | 281 | 107 | |||
| XM_015136849.2 | XP_014992335.2 | 283 | 107 | |||
| XM_015136850.2 | XP_014992336.2 | 321 | 107 | |||
| GenBank mRNA | JU333519.1 | - | 223 | 107 | ||
| JU474788.1 | - | 156 | 107 | |||
More Information
Set Membership: |
|

DOP1A
Genomic Map