Assay Details
| Gene Symbol: TRIM59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131714.2 | XP_014987200.2 | 631 | 103 | ||
| XM_015131715.2 | XP_014987201.2 | 312 | 103 | |||
| XM_015131716.2 | XP_014987202.2 | 87 | 103 | |||
| GenBank mRNA | JU334115.1 | - | 190 | 103 | ||
| JU475260.1 | - | 158 | 103 | |||
| JV712940.1 | - | 144 | 103 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015131714.2, XM_015131715.2, XM_015131716.2 transcripts.
Set Membership: |
|

TRIM59
Genomic Map