Assay Details
| Gene Symbol: C20H16orf90 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001093526.4 | XP_001093526.1 | 628 | 102 | ||
| XM_015125432.2 | XP_014980918.1 | 594 | 102 | |||
| XM_028840961.1 | XP_028696794.1 | 594 | 102 | |||
| XM_028840962.1 | XP_028696795.1 | 628 | 102 | |||
| XM_028840963.1 | XP_028696796.1 | 213 | 102 | |||
| GenBank mRNA | JV590860.1 | - | 123 | 102 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_028840963.1 transcripts.
Set Membership: |
|

C20H16orf90
Genomic Map