Assay Details
| Gene Symbol: CEP20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001109724.4 | XP_001109724.3 | 298 | 131 | ||
| XM_015125697.2 | XP_014981183.1 | 298 | 131 | |||
| XM_015125698.2 | XP_014981184.1 | 298 | 131 | |||
| GenBank mRNA | JU474274.1 | - | 236 | 131 | ||
More Information
Set Membership: |
|

CEP20
Genomic Map