Assay Details
| Gene Symbol: CCN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015148950.2 | XP_015004436.2 | 1074 | 119 | ||
| XM_028828444.1 | XP_028684277.1 | 913 | 119 | |||
| GenBank mRNA | JV048591.1 | - | 638 | 119 | ||
| JV664558.1 | - | 638 | 119 | |||
More Information
Set Membership: |
|

CCN5
Genomic Map