Assay Details
| Gene Symbol: WTIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028839550.1 | XP_028695383.1 | 903 | 65 | ||
| XM_028839551.1 | XP_028695384.1 | 918 | 65 | |||
| XM_028839552.1 | XP_028695385.1 | 918 | 65 | |||
| GenBank mRNA | JV051590.1 | - | 514 | 65 | ||
| JV571074.1 | - | 470 | 65 | |||
More Information
Set Membership: |
|

WTIP
Genomic Map