Assay Details
| Gene Symbol: PDIK1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131774.2 | XP_014987260.1 | 138 | 52 | ||
| GenBank mRNA | JU320064.1 | - | 61 | 52 | ||
| JV046264.1 | - | 46 | 52 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015131774.2 transcripts.
Set Membership: |
|

PDIK1L
Genomic Map