Assay Details
| Gene Symbol: FZD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015110810.2 | XP_014966296.1 | 1786 | 83 | ||
| GenBank mRNA | JU331912.1 | - | 215 | 83 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FZD5| Gene Symbol: FZD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015110810.2 | XP_014966296.1 | 1786 | 83 | ||
| GenBank mRNA | JU331912.1 | - | 215 | 83 | ||
Set Membership: |
|