Assay Details
| Gene Symbol: ZFP37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002800007.3 | XP_002800053.3 | 336 | 104 | ||
| XM_015117056.2 | XP_014972542.2 | 339 | 104 | |||
| XM_028834969.1 | XP_028690802.1 | 311 | 104 | |||
| XM_028834970.1 | XP_028690803.1 | 308 | 104 | |||
| GenBank mRNA | JU348107.1 | - | 242 | 104 | ||
| JV722712.1 | - | 223 | 104 | |||
More Information
Set Membership: |
|

ZFP37
Genomic Map