Assay Details
| Gene Symbol: ZNF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125029.2 | XP_014980515.1 | 427 | 101 | ||
| XM_015125030.2 | XP_014980516.1 | 427 | 101 | |||
| GenBank mRNA | JU325564.1 | - | 200 | 101 | ||
| JU472093.1 | - | 412 | 101 | |||
| JV635663.1 | - | 415 | 101 | |||
More Information
Set Membership: |
|

ZNF8
Genomic Map