Assay Details
| Gene Symbol: CRIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015144626.2 | XP_015000112.1 | 743 | 90 | ||
| XM_028851961.1 | XP_028707794.1 | 743 | 90 | |||
| XM_028851962.1 | XP_028707795.1 | 1286 | 90 | |||
| XM_028851963.1 | XP_028707796.1 | 419 | 90 | |||
| XM_028851964.1 | XP_028707797.1 | 222 | 90 | |||
| GenBank mRNA | JU333006.1 | - | 564 | 90 | ||
| JU474362.1 | - | 539 | 90 | |||
| JV046234.1 | - | 566 | 90 | |||
| JV571763.1 | - | 571 | 90 | |||
More Information
Set Membership: |
|

CRIP2
Genomic Map