Assay Details
| Gene Symbol: PRCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120442.2 | XP_014975928.1 | 1576 | 111 | ||
| GenBank mRNA | JV600211.1 | - | 332 | 111 | ||
| JV639115.1 | - | 232 | 111 | |||
More Information
Set Membership: |
|

PRCD
Genomic Map