Assay Details
| Gene Symbol: CHST5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV710955.1 | - | 26 | 76 | ||
| JV710957.1 | - | 26 | 76 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHST5| Gene Symbol: CHST5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JV710955.1 | - | 26 | 76 | ||
| JV710957.1 | - | 26 | 76 | |||
Set Membership: |
|