Assay Details
| Gene Symbol: TTC9B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015123971.2 | XP_014979457.2 | 924 | 104 | ||
| GenBank mRNA | JU327101.1 | - | 486 | 104 | ||
| JV534339.1 | - | 527 | 104 | |||
| JV606796.1 | - | 551 | 104 | |||
More Information
Set Membership: |
|

TTC9B
Genomic Map