Assay Details
| Gene Symbol: LMOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015120267.2 | XP_014975753.1 | 3290 | 67 | ||
| GenBank mRNA | JV046670.1 | - | 2013 | 67 | ||
| JV636084.1 | - | 1921 | 67 | |||
More Information
Set Membership: |
|

LMOD1
Genomic Map