Assay Details
| Gene Symbol: ZNF391 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001088444.4 | XP_001088444.2 | 413 | 110 | ||
| XM_015135759.2 | XP_014991245.2 | 334 | 110 | |||
| XM_015135760.2 | XP_014991246.2 | 445 | 110 | |||
| GenBank mRNA | JV059962.1 | - | 99 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001088444.4, XM_015135759.2, XM_015135760.2 transcripts.
Set Membership: |
|

ZNF391
Genomic Map