Assay Details
| Gene Symbol: ANKRD33B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001086061.3 | XP_001086061.2 | 487 | 100 | ||
| GenBank mRNA | JV626267.1 | - | 476 | 100 | ||
| JV721481.1 | - | 447 | 100 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD33B| Gene Symbol: ANKRD33B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001086061.3 | XP_001086061.2 | 487 | 100 | ||
| GenBank mRNA | JV626267.1 | - | 476 | 100 | ||
| JV721481.1 | - | 447 | 100 | |||