Assay Details
| Gene Symbol: HARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015141039.2 | XP_014996525.2 | 837 | 69 | ||
| XM_028849756.1 | XP_028705589.1 | 800 | 69 | |||
| XM_028849757.1 | XP_028705590.1 | 837 | 69 | |||
| XM_028849758.1 | XP_028705591.1 | 962 | 69 | |||
| XM_028849759.1 | XP_028705592.1 | 932 | 69 | |||
| XM_028849760.1 | XP_028705593.1 | 860 | 69 | |||
| GenBank mRNA | JU322890.1 | - | 836 | 69 | ||
| JU322891.1 | - | 836 | 69 | |||
| JU471480.1 | - | 818 | 69 | |||
| JU471481.1 | - | 818 | 69 | |||
| JV045462.1 | - | 821 | 69 | |||
| JV045463.1 | - | 821 | 69 | |||
| JV601797.1 | - | 837 | 69 | |||
| JV667245.1 | - | 822 | 69 | |||
More Information
Set Membership: |
|

HARS2
Genomic Map