Assay Details
Gene Symbol: C1H1orf162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148206.2 | XP_015003692.1 | 138 | 75 | ||
GenBank mRNA | JU323507.1 | - | 51 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015148206.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
