Assay Details
| Gene Symbol: TRIM13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015121091.2 | XP_014976577.2 | 113 | 87 | ||
| GenBank mRNA | JU478625.1 | - | 112 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015121091.2 transcripts.
Set Membership: |
|

TRIM13
Genomic Map