Assay Details
| Gene Symbol: CDK5R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015111099.2 | XP_014966585.1 | 1533 | 103 | ||
| GenBank mRNA | JV486037.1 | - | 318 | 103 | ||
| JV632562.1 | - | 1492 | 103 | |||
| JV668233.1 | - | 1487 | 103 | |||
More Information
Set Membership: |
|

CDK5R2
Genomic Map