Assay Details
| Gene Symbol: LOC713490 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015113621.2 | XP_014969107.1 | 1519 | 112 | ||
| XM_015113622.1 | XP_014969108.1 | 1250 | 112 | |||
| XM_015113623.2 | XP_014969109.1 | 1331 | 112 | |||
| GenBank mRNA | JU320134.1 | - | 1183 | 112 | ||
| JU470922.1 | - | 1185 | 112 | |||
| JV044816.1 | - | 1172 | 112 | |||
| JV703853.1 | - | 1203 | 112 | |||
More Information
Set Membership: |
|

LOC713490
Genomic Map