Assay Details
| Gene Symbol: EXOSC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015126515.2 | XP_014982001.1 | 815 | 82 | ||
| GenBank mRNA | JU326289.1 | - | 219 | 82 | ||
| JU473632.1 | - | 577 | 82 | |||
| JV641814.1 | - | 483 | 82 | |||
More Information
Set Membership: |
|

EXOSC6
Genomic Map