Assay Details
| Gene Symbol: C2H3orf33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015131858.2 | XP_014987344.2 | 565 | 107 | ||
| XM_015131859.2 | XP_014987345.2 | 565 | 107 | |||
| XM_028843799.1 | XP_028699632.1 | 461 | 107 | |||
| XM_028843800.1 | XP_028699633.1 | 599 | 107 | |||
| GenBank mRNA | JU334099.1 | - | 367 | 107 | ||
| JV649010.1 | - | 155 | 107 | |||
More Information
Set Membership: |
|

C2H3orf33
Genomic Map